Magne HaloTag Beads, 20% Slurry, Promega
Supplier: Promega Corporation
G7282
PAG7282EA 646.78 USD
PAG7282
Magne HaloTag Beads, 20% Slurry, Promega
Beads Magnetic Beads
The Magne HaloTag Beads provide a convenient method to covalently capture HaloTag fusion proteins with magnetic particles for protein pull-downs and purification. The beads offer high binding capacity with low nonspecific protein binding.
  • Magnetic Capture of HaloTag(R) Fusion Proteins for Pull-Downs and Purification
  • Bind ≥20mg of purified HaloTag(R) fusion protein per ml of particles
  • Manual or high-throughput applications
The Magne HaloTag Beads provide a convenient method to covalently capture HaloTag fusion proteins with magnetic particles for protein pull-downs and purification. These magnetic beads offer a high binding capacity (>/=20mg/ml) for purified HaloTag fusion proteins with low nonspecific protein binding. The magnetic handling properties allow streamlined protein purification and eliminate the need for multiple centrifugation steps, facilitating automated applications on robotic platforms. The Magne HaloTag Beads (Cat.# G7281 and G7282) are the recommended replacement for the discontinued HaloLink Magnetic Beads (Cat.# G9311).