200 bp DNA Step Ladder, 100 µg, Promega
Supplier: Promega Corporation
G6961
PAG6961EA 366.81 USD
PAG6961
200 bp DNA Step Ladder, 100 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Thirty-three blunt-ended DNA fragments ranging from 200bp to 6,600bp in 200bp increments. The 1,000bp band appears more intense than all other bands.
  • Thirty-Three Blunt-Ended DNA Fragments of 200-6,600bp in 200bp Increments
  • Load 1microl/lane
  • Supplied at a concentration of 1µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 200bp DNA Step Ladder: Thirty-three blunt-ended DNA fragments ranging from 200bp to 6,600bp in 200bp increments. The 1,000bp band appears more intense than all other bands, which are of approximately equal intensity. Recommended loading: 1microl/lane.