1 kb DNA Step Ladder, 90 µg, Promega
Supplier: Promega Corporation
G6941
PAG6941EA 239.73 USD
PAG6941
1 kb DNA Step Ladder, 90 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Ten blunt-ended DNA fragments ranging from 1kb to 10kb in 1kb increments.
  • Ten Blunt-Ended DNA Fragments of 1-10kb in 1kb Increments
  • Load 1microl/lane
  • Supplied at a concentration of 0.3µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 1kb DNA Step Ladder: Ten blunt-ended DNA fragments ranging from 1kb to 10kb in 1kb increments. All bands except the 5kb band are of equal intensity; the 5kb band is ~3 times more intense.