HaloTag Mammalian Protein Purification System, 1 each, Promega
Supplier: Promega Corporation
G6790
PAG6790EA 872.61 USD
PAG6790
HaloTag Mammalian Protein Purification System, 1 each, Promega
Protein Purification Protein Purification Kits
HaloTag Mammalian Protein Purification System is an optimized kit for purification of HaloTag fusion proteins from mammalian cell culture lysates. HaloTag Mammalian Protein Detection and Purification System also includes HaloTag TMRDirect Ligand.
  • Recover Highly Pure Recombinant Protein from Mammalian Cell Cultures, Even at Low Expression Levels
  • HaloLink™ Resin covalently binds >7mg/ml of HaloTag(R) fusion protein (10X more capacity than FLAG(R))
  • Easy to scale up or down to obtain milligram + quantities of protein
  • Choose from purification, purification + detection or sample purification + detection kits
The HaloTag Mammalian Protein Purification System (Cat.# G6790) is an optimized kit for purification of HaloTag fusion proteins from mammalian cell culture lysates. HaloTag is a protein fusion tag engineered to form a highly specific and covalent bond with the HaloLink Resin. The covalent binding coupled with the low nonspecific binding of the HaloLink Resin provides superior purity and recovery of recombinant proteins from cultured mammalian cells, even at low expression levels. The HaloTag Mammalian Protein Detection and Purification System (Cat.# G6795) also includes HaloTag TMRDirect Ligand. The simple-to-use fluorescent detection of the HaloTag fusion facilitates rapid optimization of expression and purification conditions.