Protease Inhibitor Cocktail, 50X, Promega
Supplier: Promega Corporation
G6521
PAG6521EA 109.04 USD
PAG6521
Protease Inhibitor Cocktail, 50X, Promega
Cell Culture Supplements and Additives
Protease Inhibitor Cocktail is used to prevent protein degradation after lysing cells. The product is a mixture of six different protease inhibitors with different target protease specificities.
  • Prevents Protein Degradation after Cell Lysis
  • Effective against many proteases
  • Best-in-class protease inhibition
  • Works with a wide array of fusion tags (e.g., Flag(R), His tag, GST) and capture technologies
Protease Inhibitor Cocktail is used to prevent protein degradation after lysing cells. The product is a mixture of six different protease inhibitors with different target protease specificities. The inhibitor cocktail is EDTA-free and provided as a powder, ready for reconstitution in 1ml of either 100% ethanol or DMSO to obtain a 50X working solution.