1 kb DNA Ladder, 500 µl, Promega
Supplier: Promega Corporation
G5711
PAG5711EA 214.66 USD
PAG5711
1 kb DNA Ladder, 500 µl, Promega
Electrophoresis Reagents Molecular Weight Markers
13 blunt-end fragments from 250bp to 10,000bp.
  • Thirteen Blunt-Ended Fragments of 250-10,000bp
  • Load 5microl/lane
  • Supplied at a concentration of 0.1µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Ladders are ladders with defined sizes. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. Promega 1kb Ladder: Thirteen blunt-ended fragments with sizes ranging from 250bp to 10,000bp. The 1,000bp and 3,000bp fragments have increased intensity relative to the other bands on ethidium bromide-stained agarose gels for easy identification. All other fragments are of equal intensity. The ladder is dephosphorylated and ready for 5' end-labeling with radioisotopes using T4 Polynucleotide Kinase, allowing visualization by autoradiography. Recommended loading: 5microl/lane