25 bp DNA Step Ladder, 100 µg, Promega
Supplier: Promega Corporation
G4511
PAG4511EA 276.16 USD
PAG4511
25 bp DNA Step Ladder, 100 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Twelve DNA fragments ranging from 25bp to 300bp in 25bp increments includes a visible 1,800bp "backbone" fragment.
  • Twelve DNA Fragments of 25-300bp in 25bp Increments
  • Includes 1,800bp "backbone" fragment
  • Load 5microl/lane
  • Supplied at a concentration of 0.36µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 25bp DNA Step Ladder: Twelve DNA fragments ranging from 25bp to 300bp in 25bp increments. An 1,800bp backbone fragment is also visible. The 300bp band is ~3 times more intense than all other bands. Recommended loading: 5microl/lane.