DNA Step Ladder 10 bp, 32.5 µg
Supplier: Promega Corporation
G4471
PAG4471EA 276.16 USD
PAG4471
DNA Step Ladder 10 bp, 32.5 µg
Electrophoresis Reagents Molecular Weight Markers
Ten blunt-ended DNA fragments ranging from 10bp to 100bp in exactly 10bp increments
  • Ten Blunt-Ended DNA Fragments of 10-100bp in 10bp Increments
  • Load 1microl/lane
  • Supplied at a concentration of 0.65µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
The DNA Step Ladders are ladders of defined sizes with exact incremental steps between bands. The ladders are not intended for use in quantitative analysis. Each ladder is provided with a tube of 6X Blue/Orange Loading Dye. The fragments may be stained with ethidium bromide. 10bp DNA Step Ladder: Ten blunt-ended DNA fragments ranging from 10bp to 100bp in exactly 10bp increments. All of the bands are of approximately equal intensity with the exception of the 10bp band, which may appear slightly less intense. Recommended loading: 1microl/lane.