PhiX174 DNA/HaeIII Markers, 50 µg, Promega
Supplier: Promega Corporation
G1761
PAG1761EA 312.5 USD
PAG1761
PhiX174 DNA/HaeIII Markers, 50 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
Eleven phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 72bp to 1,353bp.
  • Eleven DNA Fragments of 72-1,353bp
  • Load 1microl/lane
  • Supplied at a concentration of 1µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
Conventional DNA Digest Markers are created by digesting either lambda DNA, PhiX174 replicative form DNA, or plasmids to completion with one or more restriction enzymes. The enzymes are heat-inactivated, and the DNA fragments are either phenol-extracted, then ethanol-precipitated or just ethanol-precipitated. The precipitated fragments are resuspended in storage buffer. The markers are not intended for quantitative analysis. Each marker is supplied with a tube of 6X Blue/Orange Loading Dye. PhiX174 DNA/HaeIII Markers: Eleven phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 72bp to 1,353bp. Recommended Loading: Load 1microl/lane.