PhiX174 DNA/HinfI Markers, 50 µg, Promega
Supplier: Promega Corporation
G1751
PAG1751EA 305.7 USD
PAG1751
PhiX174 DNA/HinfI Markers, 50 µg, Promega
Electrophoresis Reagents Molecular Weight Markers
This conventional DNA digest marker is generated by ΦX174 DNA digested to completion with HinfI to create 20 ethanol-precipitated DNA fragments ranging in size from 24bp to 726bp.
  • Twenty DNA Fragments of 24-726bp
  • Load 1microl/lane
  • Supplied at a concentration of 1µg/microl
  • Supplied with 6X Blue/Orange Loading Dye
Conventional DNA Digest Markers are created by digesting either lambda DNA, PhiX174 replicative form DNA, or plasmids to completion with one or more restriction enzymes. The enzymes are heat-inactivated, and the DNA fragments are either phenol-extracted, then ethanol-precipitated or just ethanol-precipitated. The precipitated fragments are resuspended in storage buffer. The markers are not intended for quantitative analysis. Each marker is supplied with a tube of 6X Blue/Orange Loading Dye. PhiX174 DNA/HinfI Markers: Twenty ethanol-precipitated DNA fragments ranging in size from 24bp to 726bp. Recommended Loading: Load 1microl/lane.