GloMax Luminometer Light Plate, Promega
Supplier: Promega Corporation
E6531
PAE6531EA 2109.69 USD
PAE6531
GloMax Luminometer Light Plate, Promega
Microplate Readers
The GloMax® 96 Microplate Luminometer Light Plate provides a quick and easy means to verify the performance of the GloMax® 96 Microplate Luminometer. Users can check the sensitivity, reproducibility and linearity.
  • Powered by a battery
The Light Plate consists of three highly stable light sources that simulate luminescent samples at signal levels spanning four decades.