rATP, 100 mM, 400 µl, Promega
Supplier: Promega Corporation
E6011
PAE6011EA 99.88 USD
PAE6011
rATP, 100 mM, 400 µl, Promega
Nucleic Acid Reagents Nucleotides
High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.
  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400µl rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC
Ribonucleotide Triphosphates (rNTPs) are provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.