K562 Genomic DNA, 80 µg, Promega
Supplier: Promega Corporation
E4931
PAE4931EA 370.91 USD
PAE4931
K562 Genomic DNA, 80 µg, Promega
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. Supplied at a concentration of 400microg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).
  • Purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. Supplied at a concentration of 400microg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).
K562 Genomic DNA is purified from the human lymphoma cell line K562 by detergent lysis and proteinase K digestion. K562 Genomic DNA is supplied at a concentration of 400microg/ml in TE buffer (10mM Tris-HCl, 1mM EDTA; final pH 8.0).