Disposable Polypropylene Test Tubes (8×50mm), 1000/pack, Promega

Supplier: Promega Corporation
E4221
PAE4221EA 291.57 USD
PAE4221
Disposable Polypropylene Test Tubes (8×50mm), 1000/pack, Promega
Tubes Microcentrifuge Tubes
These accessories are available for the now-discontinued 20/20 luminometer.

  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20 n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued

These accessories are only for the now-discontinued Turner BioSystems Luminometer Model TD-20/20. They are not for the Turner BioSystems 20/20 n Single-Tube Luminometer or Promega GloMax 20/20 Luminometer. Turner BioSystems and Promega have teamed up to develop the next-generation luminometers for single-tube and 96-well plate analysis. For more information, see the GloMax Luminometer catalog pages or contact Technical Support.

Storage: 15 to 30 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR