pGL4.48[luc2P/SBE/Hygro] Vector, 20 µg, Promega
Supplier: Promega Corporation
E3671
PAE3671EA 545.85 USD
PAE3671
pGL4.48[luc2P/SBE/Hygro] Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
The pGL4.45[luc2P/ISRE/Hygro], pGL4.47[luc2P/SIE/Hygro], pGL4.48[luc2P/SBE/Hygro] and pGL4.52[luc2P/STAT5RE/Hygro] Vectors are optimized for expression in mammalian cells. Use these vectors to explore cytokine signaling pathways.
  • Cytokine Response Elements in Firefly Luciferase Reporter Vectors
  • Offer a range of response elements that are involved in cytokine signalling pathways
  • Each vector included an optimized firefly luciferase gene that codes a reporter protein with a one-hour half-life
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Creating a cell line with an indicator of a functional signaling pathway is useful for deciphering the components in a signaling pathway. These tools are made by inserting multiple repeats of a response element upstream of a minimal promoter (minP). We offer vectors that report the activity of a variety of cellular stress pathways using the optimized firefly luciferase gene luc2 in the pGL4 backbone. The luc2 gene is engineered to remove most cryptic transcription factor binding sites and improve mammalian expression through codon optimization. The luc2P and luc2CP and RapidResponse genes are luc2 genes appended with degradation sequences to influence the cellular half-life of the luc2 gene. The RapidResponse genes respond more rapidly to stimuli but at the expense of signal intensity. The luc2P (1-hour half-life) gene responds more rapidly than luc2 (3-hour half-life) with moderate signal intensity. These response element vectors also have a hygromycin resistance selectable marker for use either in transient transfection experiments or for selection of a stable cell line. To speed research, several pre-designed response element vectors based on the pGL4.27 Vector are available. Some of these also are available stable cell lines (GloResponse Cell Lines). The cytokine signaling pathway vectors available include interferon-alpha (INF-alpha) in pGL4.45[luc2P/ISRE/Hygro] Vector, interleukin 6 (IL-6) in pGL4.47[luc2P/SIE/Hygro] Vector, transforming growth factor beta (TGF-beta) in pGL4.48[luc2P/SBE/Hygro] Vector and interleukin 3 (IL-3) in pGL4.52[luc2P/STAT5RE/Hygro] Vector.