QuantiFluor RNA System, 1ml, Promega
Supplier: Promega Corporation
E3310
PAE3310EA 454.08 USD
PAE3310
QuantiFluor RNA System, 1ml, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The QuantiFluor RNA System contains a fluorescent RNA-binding dye that enables sensitive quantitation of small amounts of RNA in solution.
  • Sensitive Quantitation of Small Amounts of RNA in Solution
  • Requires less template RNA than other systems
  • More sensitive than absorbance methods for low-concentration samples
Sensitive quantitation of RNA is important for the success of downstream applications. The QuantiFluor RNA System contains a fluorescent RNA-binding dye that enables sensitive quantitation of small amounts of RNA in solution. Detecting and quantitating small amounts of RNA is a very important step that is used in many biological applications, particularly in molecular biology techniques.
Detecting and quantitating small amounts of RNA is an important step used in many biological applications particularly molecular biology techniques.