QuantiFluor ssDNA System, 1 ml, Promega
Supplier: Promega Corporation
E3190
PAE3190EA 618.96 USD
PAE3190
QuantiFluor ssDNA System, 1 ml, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The QuantiFluor ssDNA System contains a fluorescent dye that enables sensitive quantitation of small amounts of single-stranded (ssDNA) in solution.
  • Sensitive Quantitation of Single-Stranded DNA
  • More sensitive than NanoDrop for low-concentration samples
  • Requires less DNA than spectrophotometry
The QuantiFluor ssDNA System contains a fluorescent dye that enables sensitive quantitation of small amounts of single-stranded (ssDNA) in solution. Detecting and quantitating ssDNA is useful for a variety of research interests in molecular biology. These include studying ssDNA viruses, quantitating short synthetic ssDNA probes for site-directed mutagenesis, analysis of first-strand cDNAs and quantitating bisulfite-converted DNA to study DNA methylation.