Interface Tubing and Valves for Auto Injector System, 1 each, Promega
Supplier: Promega Corporation
E2211
PAE2211EA 294.98 USD
PAE2211
Interface Tubing and Valves for Auto Injector System, 1 each, Promega
Microplate Readers
These accessories are available for the now-discontinued 20/20 luminometer.
  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued
These accessories are only for the now-discontinued Turner BioSystems Luminometer Model TD-20/20. They are not for the Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax 20/20 Luminometer. Turner BioSystems and Promega have teamed up to develop the next-generation luminometers for single-tube and 96-well plate analysis. For more information, see the GloMax Luminometer catalog pages or contact Technical Support.