pCI-neo Mammalian Expression Vector, 20 µg, Promega
Supplier: Promega Corporation
E1841
PAE1841EA 545.85 USD
PAE1841
pCI-neo Mammalian Expression Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Contains a neomycin resistance gene and the CMV immediate-early enhancer/promoter region to promote constitutive expression of cloned DNA inserts in mammalian cells. Transfected cells can be selected with G-418.
  • Constitutively Express Cloned Insert for Transient or Stable Transfection
  • Select for stably transfected cells with neomycin phosphotransferase gene
  • Increase the steady-state level of RNA with the late SV40 polyadenylation signal approximately fivefold more than the early SV40 polyadenylation signal
  • Synthesize transcripts in vitro using the T7 RNA polymerase promoter or generate ssDNA in E. coli using the f1 origin of replication
The pCI-neo Mammalian Expression Vector carries the human cytomegalovirus (CMV) immediate-early enhancer/promoter region to promote constitutive expression of cloned DNA inserts in mammalian cells. This vector also contains the neomycin phosphotransferase gene, a selectable marker for mammalian cells. The pCI-neo Vector can be used for transient or stable expression by selecting transfected cells with the antibiotic G-418.