ProFection Mammalian Transfection System---Calcium Phosphate, 40 reactions, Promega
Supplier: Promega Corporation
E1200
PAE1200EA 268.82 USD
PAE1200
ProFection Mammalian Transfection System---Calcium Phosphate, 40 reactions, Promega
Transfection Reagents
Calcium Phosphate-mediated transfection reagent used for transfection of DNA into many cell types.
  • Calcium Phosphate-Mediated Transfection Method
  • Sufficient reagents for 40 high-efficiency transfections of cells plated in 100mm tissue culture dishes
  • High-efficiency stable or transient transfection of eukaryotic cells
The ProFection Mammalian Transfection System is a Calcium Phosphate-mediated transfection reagent. Each system contains sufficient reagents for 40 high-efficiency transfections of cells plated in 100mm tissue culture dishes. Calcium phosphate transfection is an effective method for the production of long-term stable transfectants. This method also works well for transient expression of transfected genes and can be used with most adherent cell lines.