2X Rapid Ligation Buffer, Promega
Supplier: Promega Corporation
C6711
PAC6711EA 140.2 USD
PAC6711
2X Rapid Ligation Buffer, Promega
Buffers Cloning Buffers
The LigaFast™ rapid DNA ligation system is designed for the efficient ligation of sticky-ended DNA inserts into plasmid vectors. Rapid ligation is based on the combination of T4 DNA Ligase with a unique 2X Rapid Ligation Buffer.
  • Ligation of cohesive ends in five minutes, blunt ends in 15 minutes at room temperature
  • Use with 5´, 3´ or blunt-ended DNA inserts
  • No additional buffer modifications required
  • No requirement to purify ligated DNA prior to heat-shock transformation in E. coli. ligations conducted at room temperature