Kanamycin Positive Control RNA, 1.2 kb, 5 µg, Promega
Supplier: Promega Corporation
C1381
PAC1381EA 111.36 USD
PAC1381
Kanamycin Positive Control RNA, 1.2 kb, 5 µg, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
1.2 kb Kanamycin Positive Control RNA is supplied in 10 mM Tris-HCl (pH 7.4), 1 mM EDTA.
  • Synthesis of double stranded cDNA from poly(A)+ RNA
Caution: Storage Requirements <−65 °C