Random Primers, 20 µg, Promega
Supplier: Promega Corporation
C1181
PAC1181EA
61.36
USD
PAC1181
Random Primers, 20 µg, Promega
Nucleic Acid Reagents
Primers and Probes
Use for first-strand cDNA synthesis with a reverse transcriptase.
- Streamline Universal cDNA Synthesis
- High-quality DNA hexamers of randomized sequence
Random Primers can be used for first-strand cDNA synthesis and cloning; they also are available as components of the Universal Riboclone cDNA Synthesis System (Cat.# C4360) and Reverse Transcription System (Cat.# A3500). The primers are random hexadeoxynucleotides.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)