Oligo(dT)15 Primer, 20 µg, Promega
Supplier: Promega Corporation
C1101
PAC1101EA 96.15 USD
PAC1101
Oligo(dT)15 Primer, 20 µg, Promega
Nucleic Acid Reagents Primers and Probes
Use for first-strand cDNA synthesis with a reverse transcriptase.
  • Simplify Your First-Strand cDNA Synthesis
  • Primer for reverse transcription of polyadenylated RNAs, such as mRNAs
  • Provided at 500µg/ml
Oligo(dT)15 Primer is suitable for use as a primer for first-strand cDNA synthesis with a reverse transcriptase. The primer hybridizes to the poly(A) tail of mRNA.