Maxwell RSC Instrument, Promega

Supplier: Promega Corporation
AS4500
PAAS4500EA 45298.77 USD
PAAS4500
Maxwell RSC Instrument, Promega
Automated Nucleic Acid Purification Systems
The Maxwell Rapid Sample Concentrator (RSC) Instrument is a platform for automated purification of nucleic acid from a range of sample types. Purification methods use sample lysis and binding to paramagnetic particles as the primary separation principle.

  • Automated Nucleic Acid Purification with Integrated Quantitation
  • High-quality nucleic acid purification with minimal steps and less hands-on time
  • Integrated Quantus™ Fluorometer for sample quantitation
  • Intuitive software and touch screen interface

The Maxwell Rapid Sample Concentrator (RSC) Instrument is a platform for automated purification of nucleic acid from a range of sample types. The purification methods use sample lysis and binding to paramagnetic particles as the primary separation principle. Up to 16 samples can be prepared simultaneously in 25-60 minutes, depending on sample type. The Maxwell RSC Instrument is controlled by a graphical user interface running on a tablet PC. The instrument is supplied with a Quantus Fluorometer and integrated software that allows for extracted nucleic acid quantification measurements to be captured in the run report along with sample tracking and method run data. For research use only.

Specifications:
Dimensions: (WxDxH) 330.2 x 345.2 x 299.7 mm (13 x 13.6 x 11.8 in)
Capacity: Up to 16
Molecular Weight: ~85kDa
Processing Time: 30 to 60 min (depending on sample type and method)
UV Lamp: Avg. lifetime approximately 6,000 hrs
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR