Maxwell® 16 LEV simplyRNA Purification Kits, Promega
Supplier: Promega Corporation
AS1310
PAAS1270EA 501.66 USD
PAAS1270 PAAS1280 PAAS1310
Maxwell® 16 LEV simplyRNA Purification Kits, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
This RNA purification procedure is a simple method with minimal lysate handling before automated purification on the Maxwell 16 Instrument.
  • Application: RNA Extraction Used in Many Areas of Research Included Gene Expression and Translation
  • Simple Preprocessing Steps with No Centrifugation
  • Consistent Results and Sufficient Yield for Downstream Assays
  • All Reagents and Consumables Included
  • Versatile Protocol
The Maxwell 16 LEV simplyRNA Kits are for use with the Maxwell 16 Instrument configured with the LEV High Strength Magnetic Rod and Plunger Bar Adaptor
The kit provides reagents for processing the samples and uses prefilled cartridges for purification, maximizing simplicity and convenience. The Maxwell 16 Instrument can process from one to 16 samples in under an hour.
The low elution volume is used to generate concentrated high-quality RNA suitable for use in downstream applications such as quantitative RT-PCR.
Accessories information: Maxwell 16 LEV Cartridge Rack Available Separately, One Each