Collection Plates (4-pack), Promega
Supplier: Promega Corporation
A9161
PAA9161EA 6.15 USD
PAA9161
Collection Plates (4-pack), Promega
Microplates HTS Microplates
Collection Plates are designed to use with the Wizard(R) MagneSil(R) Plasmid Purification System for rapid high-throughput isolation of plasmid DNA.
  • Use with the Wizard(R) MagneSil(R) Plasmid Purification System
  • Pack of four 96-well plates
Collection Plates are designed to use with the Wizard(R) MagneSil(R) Plasmid Purification System for rapid high-throughput isolation of plasmid DNA.