Lysis Buffer A, Promega
Supplier: Promega Corporation
A8191
PAA8191EA 130.58 USD
PAA8191
Lysis Buffer A, Promega
Buffers Extraction Buffers
Components of the Wizard Magnetic DNA Purification System for Food, which is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc.
  • Lysis Buffers Used in Protocol for Isolating DNA from Food
  • Components of the Wizard(R) Magnetic DNA Purification System for Food available as standalone buffers
Lysis Buffer A, Food, and Lysis Buffer B, Food, are comonents of the Wizard Magnetic DNA Purification System for Food. The system is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc. The DNA extracted from the food samples is suitable for use in polymerase chain reaction (PCR) testing for Genetically Modified Organism (GMO) DNA sequences.