ReadyAmp Genomic DNA Purification System, 100 reactions, Promega
Supplier: Promega Corporation
A7710
PAA7710EA 217.22 USD
PAA7710
ReadyAmp Genomic DNA Purification System, 100 reactions, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
System for purification of single-stranded DNA from whole blood or blood stains. DNA may be used directly in amplification reactions without further manipulation. No organic extraction or ethanol precipitation.
  • Single-Stranded DNA from Whole Blood or Blood Stains
  • Removes PCR inhibitors
  • Isolated DNA can be used directly in PCR
The ReadyAmp Genomic DNA Purification System yields single-stranded DNA (ssDNA) from whole blood or blood stains that may be used directly in amplification reactions without further manipulation. The process takes less than one hour and requires no organic extractions or ethanol precipitations.