Vac-Man Jr. Laboratory Vacuum Manifold, 2-sample capacity, 1 each, Promega
Supplier: Promega Corporation
A7660
PAA7660EA 24.11 USD
PAA7660
Vac-Man Jr. Laboratory Vacuum Manifold, 2-sample capacity, 1 each, Promega
Manifolds
The ideal system for rapid, effective nucleic acid purification. Each manifold comes with 2 individually controlled One-Way Luer-Lok Stopcocks; up to 2 samples can be processed at a time.
Simultaneously process up to 2 nucleic acid samples
Use with standard 1- to 2-liter sidearm flasks
Eliminate need to handle waste during nucleic acid purification