Wizard DNA Clean-Up System, 100 preps, Promega
Supplier: Promega Corporation
A7280
PAA7280EA 272.05 USD
PAA7280
Wizard DNA Clean-Up System, 100 preps, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
A simple and effective way to purify linear and circular DNA (200-50,000bp) from many molecular biology reactions.
  • Purifies DNA (200-50,000bp) from Molecular Biology Reactions
  • Results in 15 minutes or less
  • No phenol extractions or ethanol precipitations
The Wizard DNA Clean-Up System provides a simple and effective way to purify linear and circular DNA (200-50,000bp) from many molecular biology reactions. Using a quick batch-column procedure, the entire process can be completed in 15 minutes or less with no organic extractions or ethanol precipitations. DNA is eluted in water or TE buffer, ready for use. Multiple preps may be processed easily at one time with the Vac-Man Laboratory Vacuum Manifold (Cat.# A7231).