Wizard Plus Minipreps DNA Purification System, Promega®
Supplier: Promega Corporation
A7100 A7500 A7510
PAA7100EA 237.94 USD
PAA7100 PAA7500 PAA7510
Wizard Plus Minipreps DNA Purification System, Promega®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
A simple, reliable, silica-resin-based method for rapid isolation of plasmid DNA on a miniprep scale. No organic extraction or ethanol precipitation required. Extraction completed in 15 minutes.
  • Plasmid Purification from 1-10ml Cultures
  • 15-minute protocol
  • Resin-based batch column method
  • No organic extraction or ethanol precipitation
The resin-based Wizard Plus Minipreps DNA Purification System provides a simple and reliable method for rapid isolation of plasmid DNA. When using the standard protocol, the entire miniprep process can be completed in 15 minutes or less, with no organic extractions or ethanol precipitations. Minipreps may be processed individually or in multiples with the Vac-Man (20-sample capacity, Cat.# A7231) or Vac-Man Jr. (2-sample capacity, Cat.# A7660) Laboratory Vacuum Manifold. DNA is eluted from the Wizard Minicolumn in Nuclease-Free Water (Cat.# P1193). The purified plasmid can be used directly for automated fluorescent DNA sequencing and restriction digestion without further manipulation and also can be used for in vitro transcription reactions supplemented with a ribonuclease inhibitor, such as Recombinant RNasin Ribonuclease Inhibitor (Cat.# N2511). The Wizard Minipreps DNA Purification Resin is used in the isolation and preparation of plasmid DNA in conjunction with the Wizard Plus Minipreps DNA Purification Systems. The resin is available with the systems and as a standalone product.