Wizard SV 96 PCR Clean-Up Start-Up Kit, 110 V Electrical (8×96 Preps, Manifold and Free Vacuum Pump), Promega
Supplier: Promega Corporation
A6790
PAA6790EA 2073.15 USD
PAA6790
Wizard SV 96 PCR Clean-Up Start-Up Kit, 110 V Electrical (8×96 Preps, Manifold and Free Vacuum Pump), Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wizard SV 96 PCR Clean-Up System is designed for high-throughput purification of 100bp-10kb PCR products from excess nucleotides, primers and primer dimers.
  • Bundle of 8 Purification Kits, Vacuum Manifold and Free Vacuum Pump
  • Protocol complete in as little as 20 minutes
  • Purifies 100bp to 10kb PCR products
The Wizard SV 96 PCR Clean-Up System is designed for high-throughput purification of 100bp to 10kb PCR products from excess nucleotides, primers and primer dimers. This membrane-based system allows recovery of >90% in as little as 20 minutes. The purified DNA can be used for automated fluorescent sequencing, cloning, labeling, restriction digestion or microarray analysis without further manipulation. The Wizard SV 96 PCR Clean-Up System uses 96-well filtration without the need to disassemble the manifold. Filtrate waste is delivered directly to a vacuum trap, eliminating the need to dispose of collected waste within the manifold assembly. Protocols are available for automated instruments from Beckman Coulter and PerkinElmer.