GoTaq 2-Step RT-qPCR System, 5ml, Promega®
Supplier: Promega Corporation
A6010
PAA6010EA 722.32 USD
PAA6010
GoTaq 2-Step RT-qPCR System, 5ml, Promega®
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
GoTaq RT-qPCR Systems are reagent systems for quantitative analysis of RNA. GoTaq qPCR Master Mix is a ready-to-use, stabilized 2X formulation containing GoTaq(R) Hot Start Polymerase, MgCl2, dNTPs, BRYT Green(R) Dye and a proprietary reaction buffer.
  • Optimized Components for Robust RNA Expression Analysis
  • Integrated reagents for reverse transcription and downstream qPCR, including RNasin(R) Ribonuclease Inhibitor
  • Compatible with both fast and standard cycling methods
  • Resistant to a wide range of PCR inhibitors
The GoTaq qPCR Master Mix is optimized for fast and reproducible quantitative PCR assays using a double-stranded DNA-binding dye. The master mix contains BRYT Green dye, a novel fluorescent DNA-binding dye with minimal PCR inhibition for maximum PCR efficiency and greater fluorescence enhancement upon binding to double-stranded DNA (dsDNA) than SYBR Green I. The master mix is provided as a ready-to-use, stabilized 2X formulation and contains GoTaq Hot Start Polymerase, MgCl2, dNTPs, BRYT Green Dye and a proprietary reaction buffer. This master mix contains a low level of carboxy-X-rhodamine (CXR); a separate tube of CXR reference dye is included. The GoTaq 2-Step RT-qPCR System facilitates detection and relative quantification of RNA expression levels via a two-step RT-qPCR method using integrated components: GoScript Reverse Transcription System and GoTaq qPCR Master Mix. The GoScript Reverse Transcription System includes an optimized reaction buffer and M-MLV reverse transcriptase designed to enable efficient synthesis of first-strand cDNA in preparation for PCR amplification. The cDNA product can be added directly to downstream qPCR amplification reactions. The GoTaq 1-Step RT-qPCR System enables detection and relative quantification of RNA expression levels using a one-step RT-qPCR method, combining GoScript Reverse Transcriptase and GoTaq qPCR Master Mix in a single-step real-time amplification reaction. The GoScript RT Mix for 1-Step RT-qPCR (50X) combines optimized quantities amounts of GoScript Reverse Transcriptase, RNasin Plus RNase Inhibitor and additives to enhance single-step reactions.
Caution: Store all components at –30 to –10°C upon their arrival, and protect them from light. To use components immediately, store them at 2 to 8°C, protected from light, for up to three months.