ReliaPrep Blood gDNA Miniprep System, Promega®
Supplier: Promega Corporation
A5081 A5082
PAA5081EA 362.37 USD
PAA5081 PAA5082
ReliaPrep Blood gDNA Miniprep System, Promega®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The ReliaPrep Blood gDNA Miniprep System provides a complete, ready-to-use method for purification of gDNA from up to 200ul of blood or body fluid, consistently isolating pure, intact gDNA without the use of alcohol washes or precipitations.
  • Purifies DNA from up to 200microl Blood or Body Fluids
  • No ethanol, eliminating alcohol inhibition and carryover
  • Good recovery and purity in as little as 50microl elution
  • Consistent results from run-to-run and user-to-user
The ReliaPrep Blood gDNA Miniprep System provides a complete, ready-to-use method for purification of gDNA from up to 200ul of blood or body fluid, consistently isolating pure, intact gDNA without the use of alcohol washes or precipitations. Genomic DNA can be prepared from fresh or frozen blood in less than 40 minutes with expected DNA yields of 4-10ug, depending on the white blood cell count of the blood sample.