ReliaPrep Large Volume HT gDNA Isolation System, 1 each, Promega
Supplier: Promega Corporation
A2751
PAA2751EA 3222.93 USD
PAA2751
ReliaPrep Large Volume HT gDNA Isolation System, 1 each, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The ReliaPrep Large Volume HT gDNA Isolation System isolates genomic DNA (gDNA) from 1-10ml of blood in a scalable format. The chemistry eliminates tedious centrifugation steps as well as the use of hazardous chemicals.
  • Automated DNA Isolation from 1-10ml Blood
  • Effective recovery from pristine, hemolysed or frozen samples
  • Purified DNA ready for use in downstream assays or archiving
  • Chemistry scaled for each sample, reducing waste
The ReliaPrep Large Volume HT gDNA Isolation System isolates genomic DNA (gDNA) from 1-10ml of blood in a scalable format. The chemistry eliminates tedious centrifugation steps as well as the use of hazardous chemicals, which are inherent in precipitation-based chemistries. Each reagent kit provides enough reagents to process up to 96 x 10ml whole blood samples. The system has been automated on robotic liquid-handling workstations, allowing walkaway purification of genomic DNA from 1-10ml of whole blood, regardless of sample storage or shipping conditions. For low-throughput isolation of gDNA from up to 32 samples at one time, the HSM 2.0 can be used in a manual mode, where the user performs the pipetting functions. The HSM has software that controls the instrument and directs the user through the purification protocol.