Maxwell HT 96 gDNA Blood Isolation System, Promega
Supplier: Promega Corporation
A2670 A2671
PAA2670EA 372.97 USD
PAA2670 PAA2671
Maxwell HT 96 gDNA Blood Isolation System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Maxwell HT 96 gDNA Blood Isolation System provides a simple and reliable method for the rapid isolation of gDNA in a multiwell format. The purified gDNA can be used directly in PCR assays, microarrays and next-generation sequencing applications.
  • High-Throughput DNA Extraction in 24-or 96-Well Format
  • Robust, precipitation-free protocol, No chance of "lost pellets".
  • High yields of pure DNA from pristine and challenged or hemolysed samples.
  • Pure gDNA ready for demanding applications; samples in solution; no resuspension required
The Maxwell HT 96 gDNA Blood Isolation System provides a simple and reliable method for the rapid isolation of gDNA in a multiwell format. gDNA may be purified from blood and Oragene-Discovery sample collection devices. The purified gDNA can be used directly in PCR assays, microarrays and next-generation sequencing applications. The use of paramagnetic particles for DNA capture eliminates the need for centrifugation or vacuum manifolds, making the system suitable for full automation. In addition, the system does not require an organic solvent, making it safe and convenient. DNA yields of up to 12ug are expected from input blood volumes of 350ul, depending on the WBC count of the sample. Saliva samples can have variable amounts of gDNA, and up to 18ug or more of DNA may be recovered from a 700ul Oragene collection device sample.