EDTA 10 mM, (pH 8.0), Promega
Supplier: Promega Corporation
A2631
PAA2631EA 26.3 USD
PAA2631
EDTA 10 mM, (pH 8.0), Promega
Buffers Cloning Buffers
EDTA is a chelator of divalent cations suitable for biochemistry and molecular biology applications.
  • Available in 0.5M or 10mM concentrations
  • 10mM concentration also provided as part of Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)