Vac-Man 96 Vacuum Manifold, 1 each, Promega
Supplier: Promega Corporation
A2291
PAA2291EA 517.55 USD
PAA2291
Vac-Man 96 Vacuum Manifold, 1 each, Promega
Manifolds
The Vac-Man 96 Vacuum Manifold is designed for manual use with the Promega SV 96 and SV 9600 Systems for isolating up to 96 samples simultaneously.
Eliminates the need to handle waste during nucleic acid purification
Adapts for use alone or with robotic nucleic acid purification platforms
Includes Vacuum Manifold Base, Manifold Collar and Manifold Bed