Wizard SV 9600 Plasmid DNA Purification System, Promega
Supplier: Promega Corporation
A2258
PAA2258EA 11085.54 USD
PAA2258
Wizard SV 9600 Plasmid DNA Purification System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format.
  • Silica Membrane-Based, 96-Well, High-Throughput Plasmid Purification
  • 60-minute protocol
  • Manual and automated formats
  • Spin or vacuum-based methods
The Wizard SV 96 and SV 9600 Plasmid DNA Purification Systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica-membrane, 96-well, high-throughput format. A single plate can be processed in 60 minutes or less. The purified plasmid can be used directly for automated fluorescent DNA sequencing as well as for other standard molecular biology techniques, including restriction enzyme digestion. The Wizard SV 96 and SV 9600 Systems are designed for use either in a manual format or with Beckman Coulter or PerkinElmer automated instruments.