Wizard MagneSil Sequencing Reaction Clean-Up System, Promega
Supplier: Promega Corporation
A1831 A1832 A1835
PAA1831EA 279.5 USD
PAA1831 PAA1832 PAA1835
Wizard MagneSil Sequencing Reaction Clean-Up System, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Wizard MagneSil Sequencing Reaction Clean-Up System is designed for high-throughput purification of sequencing reactions, including BigDye Terminator reactions.
  • High-Throughput Purification of DNA from Sanger Sequencing Reactions
  • Customized, walkaway methods possible
  • Purified products suitable for fluorescent sequencing
The Wizard MagneSil Sequencing Reaction Clean-Up System was developed for high-throughput purification of sequencing reactions, including BigDye Terminator reactions. Cleanup is performed using the proprietary MagneSil GREEN Paramagnetic Particles with standard, nonskirted 96-well amplification plates. No user intervention is required from the time the plates are placed on the instrument until the samples are ready for loading onto the fluorescent DNA sequencer. Protocols are available for automated instruments from Beckman Coulter and Tecan. The system relies upon the MagnaBot II for magnetic separation. The Plate Clamp 96 and Plate Stand are recommended for automated use because they ensure PCR plates are uniformly flat for liquid transfer on a robotic instrument.