Wizard Genomic DNA Purification Kit, 100 isolations × 10ml, Promega®
Supplier: Promega Corporation
A1620
PAA1620EA 1545.55 USD
PAA1620
Wizard Genomic DNA Purification Kit, 100 isolations × 10ml, Promega®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
  • Manual, Solution-Based DNA Extraction Method
  • DNA from blood, animal and plant cells, bacteria or yeast
  • 60-minute, scalable protocol
  • Kit size based on number of isolations from whole blood, other sample types may vary
The Wizard Genomic DNA Purification Kit provides a simple, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria. DNA purified with this system is suitable for a variety of applications, including amplification, digestion with restriction endonucleases and membrane hybridizations (e.g., Southern and dot/slot blots).