1,3-Dipalmitin

Supplier: BACHEM AMERICAS INC

4014008.0001
O-1200.0001BAEA 224.41 USD
O-1200.0001BA
1,3-Dipalmitin
1,3-Dipalmitin

Sequence: 1,3-Dipalmitoyl-glycerol
Synonym(s): α,γ-Dipalmitin

Formula: C₃₅H₆₈O₅
Storage Temperature: Freezer
MDL Number: MFCD00042877
CAS Number: 502-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR