Universal Wastewater Standard, NSI Lab Solutions
Supplier: SPEX CERTIPREP LLC
QCI-084
NSQCI-084CS 421 USD
NSQCI-084
Universal Wastewater Standard, NSI Lab Solutions
Standards Environmental Analysis Standards
Universal Wastewater Standard is a Ready-to-use glass bottle
  • Stable and remarkably easy to use
  • Shake and pour an aliquot
  • Parameters are derived from 1:1 Glucose:Glutamic acid at 150 mg/L each
Designed to reduce the number of standards used, this standard is certif ed for 12 conventional pollutants. Works with all listed EPA approved methods.
It is provided as a sterile solution which can be stored at room temperature prior to opening. Once opened, it should be stored tightly capped at 4°C.