Complex Nutrients Standard, NSI
Supplier: SPEX CERTIPREP LLC
QCI-064
NSQCI-064EA 99.64 USD
NSQCI-064
Complex Nutrients Standard, NSI
Standards Environmental Analysis Standards
Ready-to-use whole volume standard
Standard contains 1.50–35 mg/L of TKN and 0.50–10 mg/L of Total Phosphorus.
Includes CoA listing true value and acceptance limits. Supplied in HDPE bottle.