EPA Method 8260 Surrogate Standard, NSI
Supplier: SPEX CERTIPREP LLC
C-311
NSC-311EA 179.32 USD
NSC-311
EPA Method 8260 Surrogate Standard, NSI
Standards Environmental Analysis Standards
Mixture is packaged in methanol at a concentration of 2,500ug/ml in a 1.5ml ampule.
Mixture contains 4-bromofluorobenzene, dibromofluoromethane, and toluene-d8.