Mid-Level Solids Wastewater Standard, NSI
Supplier: SPEX CERTIPREP LLC
QCI-055
NS-QCI-055CS 337.56 USD
NS-QCI-055
Mid-Level Solids Wastewater Standard, NSI
Standards Environmental Analysis Standards
Certified for TDS, TS, and TSS
Standard contains 500–1500 mg/L of TDS, 500–1600 mg/L of TS, and 60–100 mg/L of TSS.
Includes CoA listing true value and acceptance limits.