Dggacmk

Supplier: BACHEM AMERICAS INC

4050449.0005 4050449.0025
N-1970.0005BAEA 1628 USD
N-1970.0005BA N-1970.0025BA
Dggacmk
Dggacmk

Sequence: Dansyl-Glu-Gly-Arg-chloromethylketone


Peptide and amino acid chloromethyl ketones (CMK's) are irreversible inhibitors of cysteine proteases, but they also react with serine proteases. For our amino acid CMK's (e.g. TPCK, TLCK) see the corresponding product family in the 'Amino acid' section.


Irreversible inhibitor of plasminogen activator, Factor Xa and urokinase. Dns-EGR-CMK allows direct monitoring of interactions of labeled enzyme.

Formula: C₂₆H₃₆ClN₇O₇S
Storage Temperature: Freezer
MDL Number: MFCD11975051
CAS Number: 69024-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR