Calpain Inhibitor IV
Supplier: BACHEM AMERICAS INC
4027888.0005
N-1635.0005BAEA 334.97 USD
N-1635.0005BA
Calpain Inhibitor IV
Proteins and Peptides
For additional calpain inhibitors, please see also our L-trans-Epoxysuccinyl Peptides.
Z-LLL-CHO (MG132) was shown to initiate neurite outgrowth in PC12 cells with an optimal concentration of 20 nM and to be a very strong inhibitor of 20S proteasome and of calpain.