Histidylphenylalanine

Supplier: BACHEM AMERICAS INC

4002294.0001
N-1095.0001BAEA 653.59 USD
N-1095.0001BA
Histidylphenylalanine
Histidylphenylalanine

Sequence: H-His-Phe-OH


A competitive inhibitor of histidine decarboxylase. The effect of His-Phe on gastric acid secretion has been described.

Formula: C₁₅H₁₈N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD00037858
CAS Number: 16874-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR